Learn More
Medchemexpress LLC Tranexamic acid (standard) | 1197-18-8 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0149R100MG
Encompass_Preferred
Tranexamic acid (Standard) is the analytical standard of Tranexamic acid. It is intended for research and analytical applications. Tranexamic acid (cyclocapron), a cyclic analog of lysine, is an orally active antifibrinolytic agent. It attenuates the effects of severe trauma, inhibits urokinase plasminogen activator, and ameliorates dry wrinkles. Tranexamic acid can be used for research on hemostasis.
  • Analytical standard for tranexamic acid.
  • Intended for research and analytical applications.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-50774