Learn More
Medchemexpress LLC Tranexamic acid (Standard) | 1197-18-8 | 99.9% | 50 MG
SDP
Catalog No. 5000450922
Encompass_Preferred
Tranexamic acid (Standard) is the analytical standard of Tranexamic acid, intended for research and analytical applications. Tranexamic acid (cyclocapron) is an orally active antifibrinolytic agent, a cyclic analog of lysine. It attenuates the effects of severe trauma, inhibits urokinase plasminogen activator, and ameliorates dry wrinkles. It can be used for research in hemostasis. As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used for qualitative research experiments
  • Used for quantitative research experiments
  • Used for methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-004-50922 Supplier Medchemexpress LLC Supplier No. HYB0149R50MG
Tranexamic acid (Standard) is the analytical standard of Tranexamic acid, intended for research and analytical applications. Tranexamic acid (cyclocapron) is an orally active antifibrinolytic agent, a cyclic analog of lysine. It attenuates the effects of severe trauma, inhibits urokinase plasminogen activator, and ameliorates dry wrinkles. It can be used for research in hemostasis. As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used for qualitative research experiments
  • Used for quantitative research experiments
  • Used for methodological research experiments
  • Applicable in HPLC, GC, and MS