Learn More
Expression Systems Transfection Medium, 100ml
SDP
Catalog No. NC0287714
Encompass_Preferred
Expression Systems Transfection Medium is designed to complement ESF 921 and ESF AF insect cell culture media for the cotransfection or transfection step in baculovirus vector production. Transfection Medium is a serum-free and animal-free formulation that facilitates vector DNA uptake by the insect host cells. Realize higher titers by using a formulation designed to work with ESF 921 and ESF AF.
• Serum-free, animal-free formulation works seamlessly with ESF 921 and ESF AF
• Appropriate for PEI or lipofection mediated transfection
• Optimized for use with Expres2 TR Transfection Reagent
• Increases virus vector production and transient expression
• Available in 20 mL or 100 mL
• In stock for immediate shipment
• To download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC0287714 Supplier Expression Systems Supplier No. 95020100
Expression Systems Transfection Medium is designed to complement ESF 921 and ESF AF insect cell culture media for the cotransfection or transfection step in baculovirus vector production. Transfection Medium is a serum-free and animal-free formulation that facilitates vector DNA uptake by the insect host cells. Realize higher titers by using a formulation designed to work with ESF 921 and ESF AF.
• Serum-free, animal-free formulation works seamlessly with ESF 921 and ESF AF
• Appropriate for PEI or lipofection mediated transfection
• Optimized for use with Expres2 TR Transfection Reagent
• Increases virus vector production and transient expression
• Available in 20 mL or 100 mL
• In stock for immediate shipment
• To download Instructions for Use and/or COA please visit: www.expressionsystems.com