Learn More
LAMDA BIOTECH INC Tranfection reagent: LipoFexin
Catalog No. 502045793
Encompass_Preferred
LipoFexin is a lipid-based transfection reagent that forms complex with DNA or RNA, and transports the complex into cells. It works on a variety of adherent and suspension cell lines. It can be used for transfection of either DNA or RNA into eukaryotic cells in culture media with or without serum. LipoFexin has been compared to work at the similar way and efficiency as the Lipofectamine 2000 transfection reagent. LipoFexin has the following Special Features: • Superior transfection efficiency for a broad range of cell lines. • No requirement of removal of serum from culture medium. • No requirement for washing or changing of medium after transfection. • Low cytotoxicity.
Catalog No. 50-204-5793 Supplier LAMDA BIOTECH INC Supplier No. TS310
LipoFexin is a lipid-based transfection reagent that forms complex with DNA or RNA, and transports the complex into cells. It works on a variety of adherent and suspension cell lines. It can be used for transfection of either DNA or RNA into eukaryotic cells in culture media with or without serum. LipoFexin has been compared to work at the similar way and efficiency as the Lipofectamine 2000 transfection reagent. LipoFexin has the following Special Features: • Superior transfection efficiency for a broad range of cell lines. • No requirement of removal of serum from culture medium. • No requirement for washing or changing of medium after transfection. • Low cytotoxicity.