Learn More
LAMDA BIOTECH INC Tranfection reagent: SusFexin
Catalog No. 502045795
Encompass_Preferred
SusFexin is a biodegradable polymer based transfection reagent for suspension cell transfection. When mix with DNA, it will form complex with DNA and transport the complex into a variety of suspension cell lines. A remarkable feature of the reagent is the rapid and complete degradation of polymer after transfection, leading to a much less cytotoxicity to the transfected cells and improving transfection efficiency and productivity of trans-gene expression.
Catalog No. 50-204-5795 Supplier LAMDA BIOTECH INC Supplier No. TS316
SusFexin is a biodegradable polymer based transfection reagent for suspension cell transfection. When mix with DNA, it will form complex with DNA and transport the complex into a variety of suspension cell lines. A remarkable feature of the reagent is the rapid and complete degradation of polymer after transfection, leading to a much less cytotoxicity to the transfected cells and improving transfection efficiency and productivity of trans-gene expression.