Learn More
Medchemexpress LLC HY-B0195 10mg Medchemexpress, Tranilast CAS:53902-12-8 Purity:>98%
SDP
Catalog No. 502031033
Encompass_Preferred
Medchemexpress, HY-B0195 10mg Tranilast CAS:53902-12-8 Tranilast (MK-341) acts as an anti-atopic agent. Tranilast suppresses production of prostaglandin D2 (PGD2, IC50= 0.1 mM). Tranilast sodium exhibits anti-inflammatory and immunomodulatory effects. Tranilast sodium antagonizes angiotensin II and inhibits its biological effects in vascular smooth muscle cells[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1033 Supplier Medchemexpress LLC Supplier No. HYB019510MG
Medchemexpress, HY-B0195 10mg Tranilast CAS:53902-12-8 Tranilast (MK-341) acts as an anti-atopic agent. Tranilast suppresses production of prostaglandin D2 (PGD2, IC50= 0.1 mM). Tranilast sodium exhibits anti-inflammatory and immunomodulatory effects. Tranilast sodium antagonizes angiotensin II and inhibits its biological effects in vascular smooth muscle cells[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.