Learn More
Abcam Tranilast, Anti-inflammatory agent, 50MG
Catalog No. 502707400
Encompass_Preferred
MW 327.34 g/mol, Purity >99%. Potent mast cell membrane stabilizer. Exhibits a wide range of anti-inflammatory effects. Inhibits antigen-induced release of cytokines, chemokines and proteases. Able to inhibit angiotensin II induced contractions (IC₅₀ = 36 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7400 Supplier Abcam Supplier No. AB12064350MG
MW 327.34 g/mol, Purity >99%. Potent mast cell membrane stabilizer. Exhibits a wide range of anti-inflammatory effects. Inhibits antigen-induced release of cytokines, chemokines and proteases. Able to inhibit angiotensin II induced contractions (IC₅₀ = 36 μM).
The product is subject to the following: Abcam Restricted Use Statement