Learn More
Selleck Chemical LLC Tranilast 50mg 53902-12-8 SB 252218
SDP
Supplier:  Selleck Chemical LLC S143950MG
Encompass_Preferred
Tranilast (SB 252218)¿is an antiallergic drug by inhibiting lipid mediator and cytokine release from inflammatory cells, used for the treatment of allergic disorders such as asthma, allergic rhinitis and atopic dermatitis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-789-2