Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Bis(diphenylphosphino)ethylene 96.0+%, TCI America™
SDP

Catalog No. B20031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20031G Supplier TCI America Supplier No. B20031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 983-81-3
Molecular Formula C26H22P2
Molecular Weight (g/mol) 396.41
MDL Number MFCD00003046 MFCD00063219
InChI Key NCKJIJSEWKIXAT-QURGRASLSA-N
Synonym trans-1,2-bis diphenylphosphino ethylene, e-1,2-bis diphenylphosphino ethene, ethene-1,2-diylbis diphenylphosphane, e-vinylenebis diphenylphosphine, cis-1,2-bis diphenylphosphino ethylene, trans-1,2-ethenediylbis diphenylphosphine, trans-vinylenebis diphenylphosphine, cis-vinylenebis diphenylphosphine
PubChem CID 2733288
IUPAC Name [(1E)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
SMILES C(=C/P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 396.41
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,2-Bis(diphenylphosphino)ethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.