Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Bis(tri-n-butylstannyl)ethylene, 96%

Catalog No. AAL2013003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2013003 1 g
AAL2013006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2013003 Supplier Thermo Scientific Chemicals Supplier No. L2013003
  • $169.50
    $135.65 / Each of 1

    Save $33.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14275-61-7 | C26H56Sn2 | 606.154 g/mol

trans-1,2-Bis(tri-n-butylstannyl)ethylene is used as an intermediate in organic synthesis. It is also employed in the synthesis of gamma, delta-alkynones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Bis(tri-n-butylstannyl)ethylene is used as an intermediate in organic synthesis. It is also employed in the synthesis of gamma, delta-alkynones.

Solubility
Miscible with terahydrofuran and dimethyl formamide. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14275-61-7
Molecular Formula C26H56Sn2
Molecular Weight (g/mol) 606.154
MDL Number MFCD01631299
InChI Key VNKOWRBFAJTPLS-UHFFFAOYSA-N
Synonym trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, e-1,2-bis-tri-n-butylstannyl-ethylene, tributyl 2-tributylstannyl ethenyl stannane
PubChem CID 5378370
IUPAC Name tributyl-[(E)-2-tributylstannylethenyl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC

Specifications

Density 1.14
Boiling Point 185°C (0.01 mmHg)
Refractive Index 1.502
Quantity 1 g
UN Number UN2788
Beilstein 3613803
Sensitivity Air and moisture sensitive
Solubility Information Miscible with terahydrofuran and dimethyl formamide. Slightly miscible with water.
Formula Weight 606.15
Percent Purity 96%
Chemical Name or Material trans-1,2-Bis(tri-n-butylstannyl)ethylene
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H300-H372-H312-H315-H319
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Keep cold; Store under Argon; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.