Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trans-1,2-Bis(Tributylstannyl)Ethene

Catalog No. 7316255G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-162-55G 5 g
73-162-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 73-162-55G Supplier Sigma Aldrich Supplier No. 7316255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans-1,2-Bis(tributylstannyl)ethene can be used to prepare:Polyarylenevinylene polymers having triphenylamine core moieties.Two (E)-2-(2-(thiophen-2-yl)vinyl)thiophen based copolymers through Pd-catalyzed Stille coupling reaction.Vinylstannanes by reacting with aryl halides, which in turn can be used in the synthesis of substituted pyridines.

Specifications

CAS 14275-61-7
Density 1.132 g/mL at 25°C
Molecular Formula C26H56Sn2
Linear Formula C26H56Sn2
Refractive Index n20/D 1.502
MDL Number MFCD01631299
Quantity 5 g
Molecular Weight (g/mol) 606.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.