Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Bis(tributylstannyl)ethylene 98.0+%, TCI America™
SDP

Catalog No. B19751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B19751G Supplier TCI America Supplier No. B19751G

missing translation for 'validMainframeMsg'

Specifications

CAS 14275-61-7
Color Yellow
Molecular Formula C26H56Sn2
Quantity 1 g
MDL Number MFCD01631299
UN Number 2788
Synonym trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, e-1,2-bis-tri-n-butylstannyl-ethylene, tributyl 2-tributylstannyl ethenyl stannane
InChI Key VNKOWRBFAJTPLS-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC
IUPAC Name tributyl-[(E)-2-tributylstannylethenyl]stannane
Molecular Weight (g/mol) 606.154
PubChem CID 5378370
Formula Weight 606.15
Percent Purity ≥98.0% (W)
Physical Form Liquid
Chemical Name or Material trans-1,2-Bis(tributylstannyl)ethylene
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.