Learn More
Strem, An Ascensus Company CyDTA min. 98% 25g
SDP
Catalog No. 5090108960
Encompass
CAS# 125572-95-4. 25g. trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, min. 98% CyDTA. MFCD00149243. Molecular Weight: 346.32 (364.36). Molecular Formula: (HOOCCH2)2NC6H10N(CH2COOH)2∙H2O. Color/form: white pwdr. Strem# 07-0318. http://www.strem.com/catalog/v/07-0318/
Catalog No. 50-901-08960 Supplier Strem, An Ascensus Company Supplier No. 07031825G
CAS# 125572-95-4. 25g. trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, min. 98% CyDTA. MFCD00149243. Molecular Weight: 346.32 (364.36). Molecular Formula: (HOOCCH2)2NC6H10N(CH2COOH)2∙H2O. Color/form: white pwdr. Strem# 07-0318. http://www.strem.com/catalog/v/07-0318/