Learn More
Medchemexpress LLC trans-1,2-Cyclohexanediamine | 1121-22-8 | 99.24% | 114.19 | 100 G
Supplier: Medchemexpress LLC HY41957100G
trans-1,2-Cyclohexanediamine is a biochemical reagent suitable as a biological material or organic compound for life science related research. It is exclusively for research purposes and not intended for patient use. It is also utilized for the synthesis of multidentate ligands, chiral substances, and chiral stationary phases.
- Used as a biochemical reagent
- Suitable for life science research
- Synthesizes multidentate ligands
- Synthesizes chiral substances
- Synthesizes chiral stationary phases