Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Cyclohexanedimethanol 98.0+%, TCI America™
SDP

Catalog No. C23621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23621G Supplier TCI America Supplier No. C23621G
Add to Cart
Add to Cart

Specifications

CAS 25712-33-8
Melting Point 57°C
Color White-Yellow
Boiling Point 113°C
Molecular Formula C8H16O2
Quantity 1 g
Synonym trans-1,2-Bis(hydroxymethyl)cyclohexane
InChI Key XDODWINGEHBYRT-YUMQZZPRSA-N
SMILES C1CCC(C(C1)CO)CO
IUPAC Name [(1R,2R)-2-(hydroxymethyl)cyclohexyl]methanol
Molecular Weight (g/mol) 144.214
PubChem CID 11217323
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,2-Cyclohexanedimethanol
Show More Show Less
EINECSNumber (3)-2326
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.