Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-trans-1,2-Diaminocyclohexane

Catalog No. 27001610ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-001-610ML 10 mL
27-001-650ML 50 mL
27-001-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 27-001-610ML Supplier Sigma Aldrich Supplier No. 27001610ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+/-)-trans-1,2-Diaminocyclohexane

Specifications

CAS 1121-22-8
Melting Point 14°C to 15°C (lit.)
Density 0.951 g/mL (at 25°C (literature))
Boiling Point 79°C to 81°C (15 mmHg)
Molecular Formula C6H14N2
Refractive Index n20/D 1.489 (literature)
Linear Formula C6H10(NH2)2
MDL Number MFCD00063747
Quantity 10 mL
Synonym (+/-)-trans-1,2-Cyclohexanediamine
Molecular Weight (g/mol) 114.19
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.