Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-trans-1,2-Diaminocyclohexane, 99%

Catalog No. AAL0662609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0662609 10 g
AAL0662618 50 g
AAL0662630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0662609 Supplier Thermo Scientific Chemicals Supplier No. L0662609
Add to Cart

CAS: 1121-22-8 | C6H14N2 | 114.192 g/mol

(±)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-22-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00063747
InChI Key SSJXIUAHEKJCMH-PHDIDXHHSA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

Melting Point 14°C to 15°C
Density 0.951
Boiling Point 89°C to 90°C (14 mmHg)
Flash Point 68°C (154°F)
Odor Amine-like
Refractive Index 1.489
Quantity 10 g
UN Number UN2735
Beilstein 3193807
Sensitivity Air sensitive; May form a precipitate
Solubility Information Miscible with water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material (+/-)-trans-1,2-Diaminocyclohexane
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.