Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-trans-1,2-Dibromocyclohexane 95.0+%, TCI America™
SDP

Catalog No. D241025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D241025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D241025G Supplier TCI America Supplier No. D241025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7429-37-0
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.95
MDL Number MFCD00003820
InChI Key CZNHKZKWKJNOTE-PHDIDXHHSA-N
Synonym trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane
PubChem CID 98506
IUPAC Name (1R,2R)-1,2-dibromocyclohexane
SMILES Br[C@@H]1CCCC[C@H]1Br

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 g
Formula Weight 241.95
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (+/-)-trans-1,2-Dibromocyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.