Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Dichloroethylene, MilliporeSigma™ Supelco™

Catalog No. 111020236 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0236 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-102-0236 Supplier MilliporeSigma Supelco Supplier No. 369691G
Add to Cart
Add to Cart
This item is not returnable. View return policy

trans-1,2-dichloroethylene is a chlorinated aliphatic compound, which can undergo bio-degradation by methanotrophic bacteria during simulated aquifer treatment. It can be used as an industrial solvent in the preparations of oils, fats, waxes and also in the manufacture of rubber.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 156-60-5
Vapor Pressure 5.16 psi (20°C)
Density 1.257 g/mL (at 25°C)
Flash Point 6.0°C - Closed Cup
Physical Form Neat
Linear Formula ClCH=CHCl
MDL Number MFCD00062942
Melting Point -50°C
Boiling Point 48°C
Synonym trans-1,2-Dichloroethene; trans-Acetylene dichloride
Type Analytical Standard
Quantity 1 g
Formula Weight 96.94
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.