Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene 97.0+%, TCI America™
SDP

Catalog No. B15371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15371G Supplier TCI America Supplier No. B15371G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112440-50-3
Molecular Formula C18H18N2S2
Molecular Weight (g/mol) 326.476
MDL Number MFCD00145417
InChI Key AYNDKKQQUZPETC-FOCLMDBBSA-N
Synonym 1,2-Bis(2,4,5-trimethyl-3-thienyl)-trans-1,2-dicyanoethene
PubChem CID 14122209
IUPAC Name (E)-2,3-bis(2,4,5-trimethylthiophen-3-yl)but-2-enedinitrile
SMILES CC1=C(SC(=C1C(=C(C#N)C2=C(SC(=C2C)C)C)C#N)C)C

Specifications

Melting Point 167°C
Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 326.48
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.