Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,4-Cyclohexanediol 97.0+%, TCI America™
SDP

Catalog No. C23205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23205G 5 g
C23201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23205G Supplier TCI America Supplier No. C23205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6995-79-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00063612
InChI Key VKONPUDBRVKQLM-UHFFFAOYSA-N
Synonym trans-1,4-Dihydroxycyclohexane
PubChem CID 11162
IUPAC Name cyclohexane-1,4-diol
SMILES C1CC(CCC1O)O

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 116.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,4-Cyclohexanediol
EINECSNumber (3)-2310
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.