Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1-(Boc-amino)-4-cyanocyclohexane, 97%, Thermo Scientific Chemicals

Catalog No. AAH6206003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6206003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6206003 Supplier Thermo Scientific Chemicals Supplier No. H6206003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 873537-32-7 | Molecular Formula: C12H20N2O2 | Molecular Weight: 224.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873537-32-7
Molecular Formula C12H20N2O2
Molecular Weight (g/mol) 224.304
MDL Number MFCD11707221
InChI Key QHZJRHRMFIDRFL-UHFFFAOYSA-N
Synonym tert-butyl 4-cyanocyclohexyl carbamate, tert-butyl cis-4-cyanocyclohexylcarbamate, 4-cyano-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-cyanocyclohexylcarbamate, tert-butyl n-4-cyanocyclohexyl carbamate, tert-butyl 1r,4r-4-cyanocyclohexyl carbamate, qhzjrhrmfidrfl-mgcohnpysa-n, trans-1-boc-amino-4-cyanocyclohexane, trans tert-butyl 4-cyanocyclohexyl carbamate
PubChem CID 46172880
IUPAC Name tert-butyl N-(4-cyanocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C#N

Specifications

UN Number UN3439
Quantity 1 g
Formula Weight 224.29
Percent Purity 97%
Chemical Name or Material trans-1-(Boc-amino)-4-cyanocyclohexane
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.