Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane, 97%

Catalog No. AAH6248714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6248714 25 g
AAH6248703 1 g
AAH6248706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6248714 Supplier Thermo Scientific Chemicals Supplier No. H6248714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 239074-29-4 | C12H23NO3 | 229.32 g/mol

trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.

Solubility
Slightly Soluble (4.4 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 239074-29-4
Molecular Formula C12H23NO3
Molecular Weight (g/mol) 229.32
MDL Number MFCD03844605
InChI Key SGNKPJPMWHKOJO-UHFFFAOYSA-N
Synonym tert-butyl trans-4-hydroxymethyl cyclohexylcarbamate, tert-butyl trans-4-hydroxymethyl cyclohexyl carbamate, tert-butyl cis-4-hydroxymethyl cyclohexyl carbamate, tert-butyl n-4-hydroxymethyl cyclohexyl carbamate, trans-n-4-hydroxymethyl cyclohexyl carbamic acid tert-butyl ester, tert-butyl 4-hydroxymethyl cyclohexyl carbamate, tert-butyl trans-4-hydroxymethyl-cyclohexylcarbamate, trans-tert-butyl 4-hydroxymethyl cyclohexyl carbamate, tert-butyl cis-4-hydroxymethyl cyclohexylcarbamate
PubChem CID 10513598
IUPAC Name tert-butyl N-[4-(hydroxymethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CO

Specifications

Solubility Information Slightly Soluble (4.4g/L) (25°C).
Quantity 25 g
Formula Weight 229.31
Percent Purity 97%
Physical Form Solid
Chemical Name or Material trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.