Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-1-Methoxy-3-trimethylsiloxy-1,3-butadiene

Catalog No. 2128305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-283-05G 5 g
21-283-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-283-05G Supplier Sigma Aldrich Supplier No. 2128305G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans-1-methoxy-3-trimethylsiloxy-1,3-butadiene is a funtionalized diels-alder diene. mukaiyama-michael-type addition/heterocyclization of trans-1-methoxy-3-trimethylsiloxy-1,3-butadiene (danishefskyαs diene) with 1,2-diaza-1,3-butadiene has been investigated. asymmetric hetero-diels-alder cyclization of danishefskyαs diene with benzaldehyde catalyzed by mesoporous inorganic/metalorganic hybrid materials has been reported.trans-1-methoxy-3-trimethylsiloxy-1,3-butadiene was used:
  • In the synthesis of sulfone analogues of griseofulvin (sulfogriseofulvins), 4h-1-aminopyrroles and 4,5h-pyrazoles
  • As diels-alder diene for the synthesis of pyridones and pyranones
  • As reagent employed in the mannich-michael reaction for preparation of piperidinones and enaminones

Specifications

CAS 54125-02-9
Density 0.885 g/mL (at 25°C (literature))
Boiling Point 68°C to 69°C (14 mmHg)
Molecular Formula C8H16O2Si
Refractive Index n20/D 1.454 (literature)
Linear Formula (CH3)3 SiOC(=CH2)CH=CHOCH3
MDL Number MFCD00008498
Quantity 5 g
Synonym Danishefskys diene
Molecular Weight (g/mol) 172.3
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.