Learn More
Medchemexpress LLC trans-10-Heptadeceno 1mg | 126761-43-1 | 268.43 g/mol | C17H32O2 | 1 MG
SDP
Catalog No. 5000229482
Encompass_Preferred
trans-10-Heptadecenoic acid is a C17:1 trans monounsaturated fatty acid supplied for research use. It is used as a reagent or analytical standard in studies of fatty acid metabolism, lipid biosynthesis, and microbial lipid synthesis. The material is characterized by a molecular weight of 268.43 g/mol and a reported purity of 98.0%.
  • C17:1 trans monounsaturated fatty acid.
  • Purity 98.0%.
  • Suitable for biochemical and metabolic studies.
  • Provided as a 1 mg research pack for analytical or preparative applications.
  • Liquid at room temperature; colorless to light yellow.
Catalog No. 50-002-29482 Supplier Medchemexpress LLC Supplier No. HY116037A1MG
trans-10-Heptadecenoic acid is a C17:1 trans monounsaturated fatty acid supplied for research use. It is used as a reagent or analytical standard in studies of fatty acid metabolism, lipid biosynthesis, and microbial lipid synthesis. The material is characterized by a molecular weight of 268.43 g/mol and a reported purity of 98.0%.
  • C17:1 trans monounsaturated fatty acid.
  • Purity 98.0%.
  • Suitable for biochemical and metabolic studies.
  • Provided as a 1 mg research pack for analytical or preparative applications.
  • Liquid at room temperature; colorless to light yellow.