Learn More
Cayman Chemical trans-10-hydroxy Nortriptylin
Supplier: Cayman Chemical 3666010 MG
An active metabolite of amitriptyline and nortriptyline; formed from amitriptyline and nortriptyline by CYP2D6; inhibits norepinephrine reuptake in rat cortical slices and isolated human plasma (EC50s = 160 and 176 nM, respectively)