Learn More
Cayman Chemical trans-10-hydroxy Nortriptylin
SDP
Catalog No. 502913607
Encompass_Preferred
An active metabolite of amitriptyline and nortriptyline; formed from amitriptyline and nortriptyline by CYP2D6; inhibits norepinephrine reuptake in rat cortical slices and isolated human plasma (EC50s = 160 and 176 nM, respectively)
Catalog No. 50-291-3607 Supplier Cayman Chemical Supplier No. 3666025 MG
An active metabolite of amitriptyline and nortriptyline; formed from amitriptyline and nortriptyline by CYP2D6; inhibits norepinephrine reuptake in rat cortical slices and isolated human plasma (EC50s = 160 and 176 nM, respectively)