Learn More
Apexbio Technology LLC trans-13-docosenoic acid 506-33-2 5mg
SDP
Catalog No. 5000093888
Encompass_Preferred
trans-13-docosenoic acid is a long-chain monounsaturated fatty acid It is used experimentally to study lipid metabolism pathways cellular membrane processes and lipid signaling mechanisms in biological systems trans-13-docosenoic acid also serves as an analytical standard in lipidomics studies to identify and quantify fatty acid profiles in biological samples Based on these properties trans-13-docosenoic acid holds research potential in the development of lipid-based delivery systems surfactants and topical pharmaceutical and cosmetic preparations
Catalog No. 50-000-93888 Supplier Apexbio Technology LLC Supplier No. A99675
trans-13-docosenoic acid is a long-chain monounsaturated fatty acid It is used experimentally to study lipid metabolism pathways cellular membrane processes and lipid signaling mechanisms in biological systems trans-13-docosenoic acid also serves as an analytical standard in lipidomics studies to identify and quantify fatty acid profiles in biological samples Based on these properties trans-13-docosenoic acid holds research potential in the development of lipid-based delivery systems surfactants and topical pharmaceutical and cosmetic preparations