Learn More
Medchemexpress LLC (+)-trans-(1R,2R)-U-50488 hydrochloride | 109620-49-7 | 99.1% | 10 MG
SDP
Catalog No. 5000234032
Encompass_Preferred
(+)-trans-(1R,2R)-U-50488 hydrochloride is the single-enantiomer hydrochloride salt of U-50488, a selective kappa opioid receptor agonist used for pharmacology research. The compound is supplied as a high-purity solid intended for in vitro and in vivo experimental applications; store sealed and away from moisture.
  • Selective kappa opioid receptor agonist for pharmacology research.
  • High purity, 99.1% (HPLC) reported.
  • White to light yellow solid with a stable hydrochloride salt form.
  • Molecular weight 405.79 g/mol and formula C19H27Cl3N2O.
  • Available in small research quantities with cold storage recommended for solutions.
Catalog No. 50-002-34032 Supplier Medchemexpress LLC Supplier No. HY12094910MG
(+)-trans-(1R,2R)-U-50488 hydrochloride is the single-enantiomer hydrochloride salt of U-50488, a selective kappa opioid receptor agonist used for pharmacology research. The compound is supplied as a high-purity solid intended for in vitro and in vivo experimental applications; store sealed and away from moisture.
  • Selective kappa opioid receptor agonist for pharmacology research.
  • High purity, 99.1% (HPLC) reported.
  • White to light yellow solid with a stable hydrochloride salt form.
  • Molecular weight 405.79 g/mol and formula C19H27Cl3N2O.
  • Available in small research quantities with cold storage recommended for solutions.