Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Aminocyclohexanol hydrochloride, 97+%

Catalog No. p-7026361
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2232406 5 g
AAB2232414 25 g
AAB2232422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2232406 Supplier Thermo Scientific Chemicals Supplier No. B2232406
Only null left
Add to Cart

CAS: 5456-63-3 | C6H13ClNO | 150.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5456-63-3
Molecular Formula C6H13ClNO
Molecular Weight (g/mol) 150.63
MDL Number MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963
InChI Key SEBSRZIHRZRNAH-KGZKBUQUSA-N
PubChem CID 12228412
IUPAC Name (1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl
SMILES [Cl].N[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 172°C to 175°C
Quantity 5 g
Sensitivity Hygroscopic
Synonym trans-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hydrochloride, 1r, 2r-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hcl, 1r,2r-trans-2-aminocyclohexanol hydrochloride, trans-2-hydroxycyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 2-amino-, hydrochloride, 1r,2r-rel, 1r,2r-2-aminocyclohexanolhydrochloride, 1r,2r-rel-2-aminocyclohexanol hydrochloride
Formula Weight 151.64
Percent Purity ≥98%
Chemical Name or Material trans-2-Aminocyclohexanol hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.