Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-2-butene-1,4-dicarboxylic acid | 4436-74-2 | 99.17% | 100 MG
SDP

Catalog No. 5000339250
Encompass_Preferred

Trans-2-butene-1,4-dicarboxylic acid is an endogenous metabolite. It is for research use only and not sold to patients. It is also described as a metabolite for organic acidurias that can be found in urine.

  • Endogenous metabolite
  • Metabolite for organic acidurias
  • Found in urine
  • White to off-white solid
  • Chemical formula: C6H8O4
  • Molecular weight: 144.13
  • Classified as ketone, aldehyde, and acid

Catalog No. 50-003-39250 Supplier Medchemexpress LLC Supplier No. HY128426100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trans-2-butene-1,4-dicarboxylic acid is an endogenous metabolite. It is for research use only and not sold to patients. It is also described as a metabolite for organic acidurias that can be found in urine.

  • Endogenous metabolite
  • Metabolite for organic acidurias
  • Found in urine
  • White to off-white solid
  • Chemical formula: C6H8O4
  • Molecular weight: 144.13
  • Classified as ketone, aldehyde, and acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.