Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trans-2-Chloromethylvinylboronic Acid Pinacol Ester

Catalog No. 6802495G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-024-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-024-95G Supplier Sigma Aldrich Supplier No. 6802495G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans-2-Chloromethylvinylboronic acid pinacol ester is a common reactant of Suzuki coupling reaction. It can be used to prepare:Readout reagent that helps in providing qualitative and quantitative readouts in enzyme biomarker detection assays.Boronated enynes, which on Au(I) catalyzed cycloisomerization yield various dienyl boronates.+a-Methyl-substituted boronates for the allylation of ketones to yield homoallylic tertiary alcohols.

Specifications

CAS 118722-54-6
Density 0.913 g/mL at 25°C
Molecular Formula C10H15NSi
Linear Formula (CH2CHCH2)Si(CH3)2(C5H4N)
Refractive Index n20/D 1.4603
MDL Number MFCD09265157
Quantity 5 g
Synonym (E)-2-(3-Chloro-1-propenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; E-2-Chloromethylvinylboronic acid pinacol ester
Molecular Weight (g/mol) 177.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.