Learn More
Medchemexpress LLC trans-2-Heptenal 10g
SDP
Catalog No. 5000206300
Encompass_Preferred
trans-2-Heptenal has a pungent grassy smell and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of perfumes and air fresheners
Catalog No. 50-002-06300 Supplier Medchemexpress LLC Supplier No. HYW12739810G
trans-2-Heptenal has a pungent grassy smell and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of perfumes and air fresheners