Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Heptenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A114510ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A114510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A114510ML Supplier TCI America Supplier No. A114510ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16939-73-4
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00036584
InChI Key AWCPMVVOGVEPRC-VOTSOKGWSA-N
Synonym Acetic Acid trans-2-Heptenyl Ester
PubChem CID 5363203
IUPAC Name [(E)-hept-2-enyl] acetate
SMILES CCCCC=CCOC(=O)C

Specifications

Color Colorless
Boiling Point 193°C
Quantity 10 mL
Formula Weight 156.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Heptenyl Acetate
EINECSNumber (2)-0760
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.