Learn More
Medchemexpress LLC Trans-2-hexadecenal | 22644-96-8 | 99.9% | 238.41 | 1 MG
SDP
Catalog No. 5000365245
Encompass_Preferred
(E)-2-Hexadecenal (trans-2-Hexadecenal) is an MLK3 activator that induces cytoskeletal remodeling, leading to cell rounding, detachment, and ultimately apoptosis in human and mouse cells. It activates the MLK3 signaling pathway, phosphorylating MKK4/7 and JNK, subsequently activating downstream targets of JNK. This product has potential applications in research related to sphingolipid signaling pathways.
  • MLK3 activator
  • Induces cytoskeletal remodeling
  • Leads to cell rounding, detachment, and apoptosis
  • Activates MLK3 signaling pathway
  • Phosphorylates MKK4/7 and JNK
  • Activates downstream targets of JNK
  • Potential applications in sphingolipid signaling pathway research
Catalog No. 50-003-65245 Supplier Medchemexpress LLC Supplier No. 1MGHY121683
(E)-2-Hexadecenal (trans-2-Hexadecenal) is an MLK3 activator that induces cytoskeletal remodeling, leading to cell rounding, detachment, and ultimately apoptosis in human and mouse cells. It activates the MLK3 signaling pathway, phosphorylating MKK4/7 and JNK, subsequently activating downstream targets of JNK. This product has potential applications in research related to sphingolipid signaling pathways.
  • MLK3 activator
  • Induces cytoskeletal remodeling
  • Leads to cell rounding, detachment, and apoptosis
  • Activates MLK3 signaling pathway
  • Phosphorylates MKK4/7 and JNK
  • Activates downstream targets of JNK
  • Potential applications in sphingolipid signaling pathway research