Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-2-hexadecenoyl-L-carnitine | 26145-55-1 | MFCD31657521 | 97.5% | C23H43NO4 | 10MG
SDP

Catalog No. 5000261760
Encompass_Preferred

trans-2-Hexadecenoyl-L-carnitine is a long-chain acylcarnitine and endogenous metabolite commonly detected in urine. Supplied as a research reagent, it is intended for biochemical and metabolomics studies and is for research use only, not for human therapeutic use.

  • Endogenous acylcarnitine metabolite found in urine.
  • Intended for research use only.
  • Purity 97.5%.
  • Molecular formula C23H43NO4.
  • Molecular weight 397.59 g·mol⁻1.
  • Available in small milligram pack sizes suitable for analytical work.

Catalog No. 50-002-61760 Supplier Medchemexpress LLC Supplier No. HY13387110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-2-Hexadecenoyl-L-carnitine is a long-chain acylcarnitine and endogenous metabolite commonly detected in urine. Supplied as a research reagent, it is intended for biochemical and metabolomics studies and is for research use only, not for human therapeutic use.

  • Endogenous acylcarnitine metabolite found in urine.
  • Intended for research use only.
  • Purity 97.5%.
  • Molecular formula C23H43NO4.
  • Molecular weight 397.59 g·mol⁻1.
  • Available in small milligram pack sizes suitable for analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.