Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
trans-2-Hexadecenoyl-L-carnitine, ≥95.0% (LC/MS), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg
Specifications
Specifications
| CAS | 26145-55-1 |
| Linear Formula | C23H43 NO4 |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(2 E)-1-oxo-2-hexadecen-1-yl]oxy]-1-propanaminium inner salt |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 397.59 |
| Percent Purity | ≥95.0% (LC/MS) |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction