Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-2-Hexen-1-al

Catalog No. 1326595G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-265-95G 5 g
13-265-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-265-95G Supplier Sigma Aldrich Supplier No. 1326595G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans-2-hexen-1-al was used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis
  • It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples

Specifications

CAS 6728-26-3
Density 0.846 g/mL (at 25°C (literature))
Boiling Point 47°C (17 mmHg)
Molecular Formula C6H10O
Refractive Index n20/D 1.446 (literature)
Linear Formula CH3CH2CH2CH=CHCHO
MDL Number MFCD00007008
Quantity 5 g
Molecular Weight (g/mol) 98.14
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.