Learn More
Medchemexpress LLC trans-2-hexen-1-ol | 928-95-0 | MFCD00002927 | 100.0% | 100.16 g/mol | C6H12O | 100 G
SDP
Catalog No. 5000302876
Encompass_Preferred
(E)-Hex-2-en-1-ol (trans-2-hexen-1-ol; CAS 928-95-0) is the E (trans) stereoisomer of 2-hexen-1-ol, a six-carbon unsaturated primary alcohol. It is used as an organic reagent and as a fragrance/flavor intermediate in research and formulation, with high purity suitable for analytical and synthetic work.
  • High purity suitable for analytical and synthetic applications.
  • E (trans) stereoisomer with defined stereochemistry.
  • Useful reagent for organic synthesis and mechanistic studies.
  • Commonly used as a green, fruity fragrance or flavor intermediate.
  • Soluble in common organic solvents and compatible with DMSO.
  • Available in laboratory packaging sizes for research use.
Catalog No. 50-003-02876 Supplier Medchemexpress LLC Supplier No. HYW015936100G
(E)-Hex-2-en-1-ol (trans-2-hexen-1-ol; CAS 928-95-0) is the E (trans) stereoisomer of 2-hexen-1-ol, a six-carbon unsaturated primary alcohol. It is used as an organic reagent and as a fragrance/flavor intermediate in research and formulation, with high purity suitable for analytical and synthetic work.
  • High purity suitable for analytical and synthetic applications.
  • E (trans) stereoisomer with defined stereochemistry.
  • Useful reagent for organic synthesis and mechanistic studies.
  • Commonly used as a green, fruity fragrance or flavor intermediate.
  • Soluble in common organic solvents and compatible with DMSO.
  • Available in laboratory packaging sizes for research use.