Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A087725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A087725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A087725ML Supplier TCI America Supplier No. A087725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 2497-18-9
Color Colorless
Boiling Point 165°C
Molecular Formula C8H14O2
Quantity 25 mL
MDL Number MFCD00009474
Synonym z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z
InChI Key HRHOWZHRCRZVCU-AATRIKPKSA-N
SMILES CCCC=CCOC(=O)C
IUPAC Name [(E)-hex-2-enyl] acetate
Molecular Weight (g/mol) 142.198
PubChem CID 2733294
Formula Weight 142.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexenyl Acetate
Show More Show Less
EINECSNumber (2)-0760&(2)-2533
RTECSNumber MP8425000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.