Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexenyl Hexanoate 97.0+%, TCI America™
SDP

Catalog No. C090425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C090425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C090425ML Supplier TCI America Supplier No. C090425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53398-86-0
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00036551
InChI Key UQPLEMTXCSYMEK-VQHVLOKHSA-N
Synonym Hexanoic Acid trans-2-Hexenyl Ester
PubChem CID 5352973
IUPAC Name (2E)-hex-2-en-1-yl hexanoate
SMILES CCCCCC(=O)OC\C=C\CCC

Specifications

Color Colorless
Boiling Point 79°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexenyl Hexanoate
EINECSNumber (2)-0760
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.