Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hydroxy-1-cyclohexylamine Hydrochloride 99%, Thermo Scientific™

Catalog No. AC265830025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC265830025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC265830025 Supplier Thermo Scientific Chemicals Supplier No. 265830025

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5456-63-3
Molecular Formula C6H13ClNO
Molecular Weight (g/mol) 150.63
MDL Number MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963
InChI Key SEBSRZIHRZRNAH-KGZKBUQUSA-N
Synonym trans-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hydrochloride, 1r, 2r-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hcl, 1r,2r-trans-2-aminocyclohexanol hydrochloride, trans-2-hydroxycyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 2-amino-, hydrochloride, 1r,2r-rel, 1r,2r-2-aminocyclohexanolhydrochloride, 1r,2r-rel-2-aminocyclohexanol hydrochloride
PubChem CID 12228412
IUPAC Name (1R,2R)-2-aminocyclohexan-1-ol hydrochloridyl
SMILES [Cl].N[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 173°C to 177°C
Color Beige to Brown or White
Infrared Spectrum Authentic
Quantity 2.5 g
Formula Weight 151.64
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Hydroxy-1-cyclohexylamine hydrochloride, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.