Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Methylcyclohexanol 98.0+%, TCI America™
SDP

Catalog No. M03525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03525ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03525ML Supplier TCI America Supplier No. M03525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7443-52-9
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00064168
InChI Key NDVWOBYBJYUSMF-RNFRBKRXSA-N
Synonym trans-2-methylcyclohexanol, cyclohexanol, 2-methyl-, trans, 2-methylcyclohexanol trans, cyclohexanol, 2-methyl-, 1s,2r, 2-methylcyclohexanol #, trans-2-methyl-cyclohexanol, 1r,2r-2-methylcyclohexanol, 2alpha-methylcyclohexan-1beta-ol, 1r,2r-2-methyl-cyclohexanol
PubChem CID 24004
IUPAC Name (1R,2R)-2-methylcyclohexan-1-ol
SMILES CC1CCCCC1O

Specifications

Melting Point -21°C
Color Colorless
Boiling Point 167°C
Quantity 5 mL
UN Number 2617
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Methylcyclohexanol
EINECSNumber (3)-2301
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.