Learn More
Medchemexpress LLC Trans-2-nonenal | 18829-56-6 | MFCD00007012 | 140.22 g/mol | C9H16O | 500 MG
SDP
Catalog No. 5000208753
Encompass_Preferred
Analytical standard of trans-2-nonenal supplied as a liquid reference material for use in chromatographic and mass spectrometry methods. Intended for qualitative and quantitative research and method development.
  • Used as a reference standard in HPLC, GC, and MS.
  • Provided in a 500 mg package for lab-scale analyses.
  • Liquid form suitable for standard preparation and spiking experiments.
  • Certificate of analysis and safety data sheet available for handling and storage details.
Catalog No. 50-002-08753 Supplier Medchemexpress LLC Supplier No. HYW089800R500MG
Analytical standard of trans-2-nonenal supplied as a liquid reference material for use in chromatographic and mass spectrometry methods. Intended for qualitative and quantitative research and method development.
  • Used as a reference standard in HPLC, GC, and MS.
  • Provided in a 500 mg package for lab-scale analyses.
  • Liquid form suitable for standard preparation and spiking experiments.
  • Certificate of analysis and safety data sheet available for handling and storage details.