Learn More
Medchemexpress LLC Trans-2-nonenal | 18829-56-6 | MFCD00007012 | 98.8% | 140.23 g/mol | C9H16O | 250 MG
SDP
Catalog No. 5000208832
Encompass_Preferred
Trans-2-nonenal is an analytical reference standard of trans-2-nonenal (CAS 18829-56-6) supplied for research and analytical applications such as GC, HPLC, and MS. The compound is provided in small pack sizes suitable for reference and method development; a certificate of analysis and safety documentation are available.
  • analytical standard for research and analytical applications.
  • high purity, typically about 98.8% (per COA).
  • suitable for GC, HPLC, and MS method development and quantitation.
  • available in small pack sizes, including 250 MG.
  • chemical formula C9H16O and molecular weight ~140.23 g/mol.
  • safety data sheet and certificate of analysis available.
Catalog No. 50-002-08832 Supplier Medchemexpress LLC Supplier No. 250MGHYW089800R
Trans-2-nonenal is an analytical reference standard of trans-2-nonenal (CAS 18829-56-6) supplied for research and analytical applications such as GC, HPLC, and MS. The compound is provided in small pack sizes suitable for reference and method development; a certificate of analysis and safety documentation are available.
  • analytical standard for research and analytical applications.
  • high purity, typically about 98.8% (per COA).
  • suitable for GC, HPLC, and MS method development and quantitation.
  • available in small pack sizes, including 250 MG.
  • chemical formula C9H16O and molecular weight ~140.23 g/mol.
  • safety data sheet and certificate of analysis available.