Learn More
Medchemexpress LLC Trans-2-octen-1-ol 10g
SDP
Catalog No. 5000205406
Encompass_Preferred
Trans-2-octen-1-ol is an organic compound belonging to alcohols It has a strong fruity aroma and is commonly found in a variety of foods including fruits and vegetables Trans-2-octen-1-ol has various applications in the food industry especially as a flavoring agent which helps to impart a characteristic aroma and taste to certain foods In addition it has potential use in inhibiting inflammatory diseases and cancer
Catalog No. 50-002-05406 Supplier Medchemexpress LLC Supplier No. HYW09972410G
Trans-2-octen-1-ol is an organic compound belonging to alcohols It has a strong fruity aroma and is commonly found in a variety of foods including fruits and vegetables Trans-2-octen-1-ol has various applications in the food industry especially as a flavoring agent which helps to impart a characteristic aroma and taste to certain foods In addition it has potential use in inhibiting inflammatory diseases and cancer