Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-2-Octenal

Catalog No. 2699565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-995-65G 5 g
26-995-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-995-65G Supplier Sigma Aldrich Supplier No. 2699565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The rate constant for the gas-phase reaction of the no(3) radical with trans-2-octenal was studied using absolute rate method
  • The fungal volatile organic compound trans-2-octenal, caused locomotory defects and changes in green fluorescent protein (gfp) and antigen-labeled dopaminergic neurons in adult drosophila melanogaster

Specifications

CAS 2548-87-0
Density 0.846 g/mL (at 25°C (literature))
Boiling Point 84°C to 86°C (19 mmHg)
Molecular Formula C8H14O
Refractive Index n20/D 1.45 (literature)
Linear Formula CH3(CH2)4 CH=CHCHO
MDL Number MFCD00007011
Quantity 5 g
Molecular Weight (g/mol) 126.2
Percent Purity 94%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.