Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Octenoic Acid 98.0+%, TCI America™
SDP

Catalog No. O000410ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O000410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O000410ML Supplier TCI America Supplier No. O000410ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1871-67-6
Molecular Formula C8H13O2
Molecular Weight (g/mol) 141.19
MDL Number MFCD00002706
InChI Key CWMPPVPFLSZGCY-VOTSOKGWSA-M
Synonym trans-2-octenoic acid, 2-octenoic acid, e-2-octenoic acid, e-oct-2-enoic acid, 2-octenoic acid, 2e, 2e-octenoic acid, 2-octenoic acid, e, trans-alpha-octenoic acid, unii-c0057tj59e, oct-2-enoic acid
PubChem CID 5282714
ChEBI CHEBI:86544
IUPAC Name (2E)-oct-2-enoate
SMILES CCCCC\C=C\C([O-])=O

Specifications

Color Yellow
Boiling Point 143°C
Quantity 10 mL
UN Number 3265
Formula Weight 142.20
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material trans-2-Octenoic Acid
EINECSNumber (2)-0609
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.