Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-2-Penten-1-ol

Catalog No. 4244475ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-444-75ML 5 mL
42-444-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-444-75ML Supplier Sigma Aldrich Supplier No. 4244475ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans-2-penten-1-ol is an allyl alcohol. it is one of the volatile compounds found in olive oil, cashew apple juice and fermented cucumber brines. the rate constants and product ion distributions of its reaction with h3o+, no+ and o2.+ ions have been studied using selected ion flow tube (sift).trans-2-penten-1-ol may be used in the synthesis of the following:
  • Leustroducsin b
  • Trichloroacetimidate
  • (e)-2,3,3'-trifluoro-4-(2-(trans-4-pentylcyclohexyl)ethyl)-4'-(pent-2-enyloxy)biphenyl
  • Trans-1-bromo-2-pentene
  • Trans-1-chloro-2-pentene

Specifications

CAS 1576-96-1
Density 0.847 g/mL (at 25°C (literature))
Boiling Point 139°C to 139.5°C (lit.)
Molecular Formula C5H10O
Refractive Index n20/D 1.434 (literature)
Linear Formula C2H5CH=CHCH2OH
MDL Number MFCD00002926
Quantity 5 mL
Molecular Weight (g/mol) 86.13
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.