Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Pentenal, 80-85%, Thermo Scientific™

Catalog No. AC233140050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC233140050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC233140050 Supplier Thermo Scientific Chemicals Supplier No. 233140050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-87-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
InChI Key DTCCTIQRPGSLPT-ONEGZZNKSA-N
Synonym trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al
PubChem CID 5364752
IUPAC Name (E)-pent-2-enal
SMILES CCC=CC=O

Specifications

Density 0.86g/mL
Assay Percent Range 80-85%
Linear Formula CHOHC=CHC2H5
Quantity 5 g
Beilstein 01, IV, 3459
Formula Weight 84.12
Chemical Name or Material trans-2-Pentenal, 80-85%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.